Tissue processing/embedding cassettes, Histosette® II, M492
Supplier: Simport Scientific
M492-2 M492-4 M492-5 M492-6 M492-10 M492-11 M492-12 M492-3 M492-8 M492-9 M492-7
SIMPM492-2EA 0 SGD
SIMPM492-2 SIMPM492-4 SIMPM492-5 SIMPM492-6 SIMPM492-10 SIMPM492-11 SIMPM492-12 SIMPM492-3 SIMPM492-8 SIMPM492-9 SIMPM492-7
Tissue processing/embedding cassettes, Histosette® II, M492
Embedding Cassettes Tissue Processing Cassettes
Acetal polymer.
  • Resistant to the chemical action of histological solvents
  • Flow-through slots maximise fluid exchange
  • Integral lid
  • Anterior writing area at 45° angle to enable use with leading cassette labelling instruments
Order Now
SPECIFICATIONS
Environmentally Preferable