Storage Drawer
Supplier: Simport Scientific
M495-7
SIMPM495-7EA 273 SGD
SIMPM495-7
Storage Drawer
Boxes Storage Boxes
Durable, waterproof, heavy-duty cardboard.
  • Requires minimal space
  • Label provided for each box allows recording of specimen number, date and type of specimen
  • Each box can store up to 165 embedding rings or 250 cassettes