Tissue cassettes in QuickLoad™ taped stacks, Unisette™, M505T
Supplier: Simport Scientific
M505-10T M505-11T M505-12T M505-2T M505-3T M505-4T M505-5T M505-6T M505-7T M505-8T M505-9T
SIMPM505-10TEA 0 SGD
SIMPM505-10T SIMPM505-11T SIMPM505-12T SIMPM505-2T SIMPM505-3T SIMPM505-4T SIMPM505-5T SIMPM505-6T SIMPM505-7T SIMPM505-8T SIMPM505-9T
Tissue cassettes in QuickLoad™ taped stacks, Unisette™, M505T
Embedding Cassettes Tissue Processing Cassettes
Moulded from a special high density acetal polymer, these cassettes keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents.
  • M506 series is similar to M505 but specially designed to hold biopsy specimens during the embedding process
  • Available in non cytotoxic, non metallic colours
  • The snap-latch and hinge-lock design prevents early separation of base and lid and allows for one handed operation
The efficient flow-through slots maximise fluid exchange and ensure proper drainage. The one-pieceMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable