Tissue cassettes in QuickLoad™ stacks, Slimsette™, M509T
Supplier: Simport Scientific
M509-10T M509-11T M509-12T M509-2T M509-3T M509-4T M509-5T M509-6T M509-7T M509-8T M509-9T
SIMPM509-10TEA 0 SGD
SIMPM509-10T SIMPM509-11T SIMPM509-12T SIMPM509-2T SIMPM509-3T SIMPM509-4T SIMPM509-5T SIMPM509-6T SIMPM509-7T SIMPM509-8T SIMPM509-9T
Tissue cassettes in QuickLoad™ stacks, Slimsette™, M509T
Embedding Cassettes Tissue Processing Cassettes
To be used with Leica and Sakura inkjet printers.
  • The one-piece disposable plastic cover is pre-installed on each cassette
  • Eliminates the need for reusable steel lids
  • The cover can be opened and closed as often as necessary and will always relock
Moulded from a special high density acetal polymer, they keep specimens safely submerged and are totally resistant to the chemical action of solvents used in histology laboratories. The efficient flow-through slots maximise fluid exchange and ensure proper reagent drainage. The cover does not protrude aboveMore Product Information