Tissue cassettes in QuickLoad™ sleeves, Slimsette™, M509SL
Supplier: Simport Scientific
M509-10SL M509-11SL M509-12SL M509-2SL M509-3SL M509-4SL M509-5SL M509-6SL M509-7SL M509-8SL M509-9SL
SIMPM509-10SLEA 0 SGD
SIMPM509-10SL SIMPM509-11SL SIMPM509-12SL SIMPM509-2SL SIMPM509-3SL SIMPM509-4SL SIMPM509-5SL SIMPM509-6SL SIMPM509-7SL SIMPM509-8SL SIMPM509-9SL
Tissue cassettes in QuickLoad™ sleeves, Slimsette™, M509SL
Embedding Cassettes Tissue Processing Cassettes
Acetal polymer. This model is similar to the M507 cassette but is intended for tissue processing/embedding procedures.
  • Large labelling areas are located on three sides of the cassette
  • Anterior writing area at a 45° angle
  • Available in several non cytotoxic, non metallic colours
The transparent sleeve allows viewing of cassettes in order to confirm there is no jam in the sleeve during the printing process. The efficient flow-through slots maximise fluid exchange and ensure proper drainage. Lids can be opened and closed as often as necessary and theyMore Product Information