Biopsy cassettes in QuickLoad™ stacks, Slimsette™, M510T
Supplier: Simport Scientific
M510-10T M510-11T M510-12T M510-2T M510-3T M510-4T M510-5T M510-6T M510-7T M510-8T M510-9T
SIMPM510-10TEA 0 SGD
SIMPM510-10T SIMPM510-11T SIMPM510-12T SIMPM510-2T SIMPM510-3T SIMPM510-4T SIMPM510-5T SIMPM510-6T SIMPM510-7T SIMPM510-8T SIMPM510-9T
Biopsy cassettes in QuickLoad™ stacks, Slimsette™, M510T
Embedding Cassettes Biopsy Processing Cassettes
Made of acetal. Slimsette™ biopsy cassettes are similar to M509 tissue cassettes but are specially designed to hold biopsy specimens.
  • The anterior printing area is at a 45° angle and offers an unobstructed view of the writing surface
  • Available in several colours
They have an attached lid that opens from the back of the cassette. The lids arrive closed but can be opened many times, always relocking securely.
Packaging: Cassettes are packed in stacks of 40. Each case contains 50 stacks for a total of 2000 cassettes.