Tissue and biopsy cassettes in QuickLoad™ stacks, Swingsette™, M517T
Supplier: Simport Scientific
M517-4T M517-6T M517-2T M517-10T M517-11T M517-12T M517-3T M517-5T M517-7T M517-8T M517-9T
SIMPM517-4TEA 0 SGD
SIMPM517-4T SIMPM517-6T SIMPM517-2T SIMPM517-10T SIMPM517-11T SIMPM517-12T SIMPM517-3T SIMPM517-5T SIMPM517-7T SIMPM517-8T SIMPM517-9T
Tissue and biopsy cassettes in QuickLoad™ stacks, Swingsette™, M517T
Embedding Cassettes Tissue Processing Cassettes
These disposable plastic cassettes hold tissue specimens during processing and embedding, as well as in storage. Suitable for use with Leica and Sakura labelers.
  • Made of acetal
  • Taped cassettes to be used with Leica and Sakura Ink Jet printers
  • Anterior writing area at a 45° angle
  • Large tab for convenient and easy opening
  • Covers packaged separately
  • Easy-to-assemble disposable cover with special hinge
Order Now
SPECIFICATIONS
Environmentally Preferable