Slide holder, SlideTray™
Supplier: Simport Scientific
M755-20W
SIMPM755-20WEA 0 SGD
SIMPM755-20W
Slide holder, SlideTray™
Slide Holders
This drawer is made from heat resistant injection moulded plastic.
  • Handy slide removal
  • Stackable to save space