Slide staining system, StainTray™ M918
Supplier: Simport Scientific
M918-1 M918-2 M919-1 M919-2
SIMPM918-1EA 0 SGD
SIMPM918-1 SIMPM918-2 SIMPM919-1 SIMPM919-2
Slide staining system, StainTray™ M918
Staining Boxes
StainTray™ M918 offers a user friendly approach to immunohistochemistry staining.
Suitable, not only for routine staining requiring a humid chamber, but also for haematology, cytology and microbiology laboratories.
Order Now
SPECIFICATIONS
Environmentally Preferable