Slide staining system, StainTray™ M922
Supplier: Simport Scientific
M922-1 M922-2 M923-1 M923-2
SIMPM922-1EA 0 SGD
SIMPM922-1 SIMPM922-2 SIMPM923-1 SIMPM923-2
Slide staining system, StainTray™ M922
Staining Boxes
StainTray M922 is a high-capacity slide staining system.
  • Rails covered with a special 'tacky' polymer strip that holds slides in place
  • Base will hold excess stain solutions dripping from the slides
  • Drain plug can be removed to empty the StainTray from reagents
  • Stackable and four rubber feet ensures greater base stability
Suitable for hematology, cytology, and microbiology laboratories.
Order Now
SPECIFICATIONS
Environmentally Preferable