Specimen container, prefilled with 10% neutral buffered formalin, HistoTainer™
Supplier: Simport Scientific
M961-120FW
SIMPM961-120FWEA 0 SGD
SIMPM961-120FW
Specimen container, prefilled with 10% neutral buffered formalin, HistoTainer™
Sample Containers General Purpose Containers
The HistoTainer™ is half filled with 10% neutral buffered formalin as a fixative, this penetrates quickly, but fixes slowly. The formalin is enhanced by a buffering capacity optimising histological results by light microscopy and immunohistochemistry.
  • Containers are made of PP
  • Closures are made of PE
  • Container lids are ribbed for easy opening
  • Stackable for easy, safe storage and translucent to allow specimens to be viewed without opening
  • Vertical walls offer excellent rigidity
Both containers and caps are manufactured without the use of plasticisersMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable