Test tube racks, MultiRack™
Supplier: Simport Scientific
S600-13B S600-13G S600-13L S600-13O S600-13Y S600-16B S600-16G S600-16L S600-16O S600-16Y S600-30B S600-30G S600-30Y S600-30L S600-30O
SIMPS600-13BEA 0 SGD
SIMPS600-13B SIMPS600-13G SIMPS600-13L SIMPS600-13O SIMPS600-13Y SIMPS600-16B SIMPS600-16G SIMPS600-16L SIMPS600-16O SIMPS600-16Y SIMPS600-30B SIMPS600-30G SIMPS600-30Y SIMPS600-30L SIMPS600-30O
Test tube racks, MultiRack™
Racks Tube Racks
The MultiRack™ is available in three models to accommodate a full range of laboratory test tubes and centrifuge tubes up to 30 mm in diameter. Made of highly resistant acetal, as it will not shatter or stain in contact with most laboratory chemicals. These have three-tiers that facilitate the insertion and stability of tubes. The base tier has rounded wells with drain holes. Convenient handles on each side of the rack will ensure a safe grip when carrying it around.
  • Maintain stability without tipping over
  • Available in five diffferent colors
  • Compact,
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable