Deep well plates, 96-well, Bioblock™
Supplier: Simport Scientific
T110-5
SIMPT110-5EA 0 SGD
SIMPT110-5
Deep well plates, 96-well, Bioblock™
Microplates Deep Well Microplates
PP. The 1,2 ml capacity round bottom deep well plate (1 ml when capped) is easy to empty completely and ideal for culturing bacterial cells. It can withstand centrifugation up to 6000 ×g and is DMSO resistant.
Order Now
SPECIFICATIONS
Environmentally Preferable