Deep well plates, 96-well, Bioblock™
Supplier: Simport Scientific
T110-6
SIMPT110-6EA 0 SGD
SIMPT110-6
Deep well plates, 96-well, Bioblock™
Microplates Deep Well Microplates
PS. The 1,2 ml capacity round bottom deep well plate (1 ml when capped) can withstand centrifugation up to 3000×g. It is not DMSO resistant.
Delivery information: Packed in bags of 4 plates.