Cryogenic vials, Cryovial®
Supplier: Simport Scientific
T309-1A T309-2 T309-2A T309-3A T309-4A T309-5A T310-10A T310-1A T310-2 T310-3A T310-4A T311-1 T311-2 T311-3 T311-4 T311-4A T311-5 T310-5A T310-2A T308-3A T301-1 T308-2A
SIMPT309-1AEA 0 SGD
SIMPT309-1A SIMPT309-2 SIMPT309-2A SIMPT309-3A SIMPT309-4A SIMPT309-5A SIMPT310-10A SIMPT310-1A SIMPT310-2 SIMPT310-3A SIMPT310-4A SIMPT311-1 SIMPT311-2 SIMPT311-3 SIMPT311-4 SIMPT311-4A SIMPT311-5 SIMPT310-5A SIMPT310-2A SIMPT308-3A SIMPT301-1 SIMPT308-2A
Cryogenic vials, Cryovial®
Vials
PP vial and closure. Designed for the storage of biological material, human or animal cells, at temperatures as low as −196 °C (but should be used only in the gas phase of liquid nitrogen).
  • Certified RNase-, DNase-, pyrogen- and DNA-free
  • With graduations and marking area
Round bottom vials can be centrifuged up to 17000×g
Round bottom vials can be centrifuged up to 17 000 ×g
Order Now
SPECIFICATIONS
Environmentally Preferable