Cryostorage boxes, Cryostore™
Supplier: Simport Scientific
T314-581B T314-225B T314-542B T314-542G T314-542R T314-542Y
SIMPT314-581BEA 0 SGD
SIMPT314-581B SIMPT314-225B SIMPT314-542B SIMPT314-542G SIMPT314-542R SIMPT314-542Y
Cryostorage boxes, Cryostore™
Boxes Cryoboxes
PC, coloured.
  • Writable surfaces
  • Available with four colours
  • Printed with squared numbers
  • Transparent cover allows to see the content of the box
These durable cryogenic storage boxes are designed to be used at temperatures between −196 °C and +121 °C. It can accommodate around 42×T310-10A Cryovials®.
Order Now
SPECIFICATIONS
Environmentally Preferable