PCR plates, low profile
Supplier: Simport Scientific
T323-96LPN
SIMPT323-96LPNEA 0 SGD
SIMPT323-96LPN
PCR plates, low profile
Microplates PCR Plates
PP. Thin walled 96-well PCR plates with small tube volume (100 µl).
  • Alphanumeric grid for easy sample identification
  • Flat surface for better sealing
  • Can be cut to desired format
  • Small volume reducing dead space between sample and cover
Packaging: Packed in tamperproof, resealable bags of 10 plates.
Order Now
SPECIFICATIONS
Environmentally Preferable