PCR plate supports, Combi Box™ and Combi Rack™
Supplier: Simport Scientific
T327 T327-1
SIMPT327EA 0 SGD
SIMPT327 SIMPT327-1
PCR plate supports, Combi Box™ and Combi Rack™
Racks Tube Racks
Combi-Box™ PVC, Combi-Rack™ PP.
The Combi-Box™ can be used not only as a storage rack but also as a workstation. The white base will accept all 96- and 384-well plates.
Transparent cover, simple to remove, allows easy viewing of the contents
Height 40 mm, stackable
The Combi-Rack® can support up to 96 PCR tubes or 12 strips of eight tubes with caps. For storage, the Combi-Rack™ fits directly into the Combi-Box™.
Alphanumeric labelling for easy sample identification
Can be placed directly on the laboratory bench, freezer shelf, or in the refrigerator
Temperature resistant from -80 to +121 °C, autoclavable
Order Now
SPECIFICATIONS
Environmentally Preferable