Cryoboxes, PS
Supplier: Simport Scientific
T330-64
SIMPT330-64EA 0 SGD
SIMPT330-64
Cryoboxes, PS
Boxes Cryoboxes
PS, white/transparent with blue polyurethane foam insert.
  • Boxes have a usable range of –90 to +80 °C
  • Boxes are stackable for space saving in freezers, refrigerators and lab counters
  • Being printed with a series of squares (numbered 1 to 64)
Ideal for holding up to 64 tubes from 0,5 to 2 ml. A transparent cover allows the user to see the contents and is aligned in a specific orientation. The surface accepts writing with markers for better inventory control.