CapLock™ clips
Supplier: Simport Scientific
T330-75CLB T330-75CLG T330-75CLR T330-75CLW T330-75CLY
SIMPT330-75CLBEA 0 SGD
SIMPT330-75CLB SIMPT330-75CLG SIMPT330-75CLR SIMPT330-75CLW SIMPT330-75CLY
CapLock™ clips
Clamps General Purpose Clamps
These CapLock™ clips are made of PE. They provide extra security against inadvertent cap openings on critical samples or during incubation above 80 °C.
  • Ideal for shipping samples
  • Suitable for 5 ml tubes with snap cap