Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Supplier: Simport Scientific
T332-2 T332-2S T332-4 T332-4S T332-5 T332-5S T332-6 T332-6S T332-6SPR T332-7 T332-7S T332-2SPR T332-4SPR T332-5SPR T332-7SPR
SIMPT332-2EA 0 SGD
SIMPT332-2 SIMPT332-2S SIMPT332-4 SIMPT332-4S SIMPT332-5 SIMPT332-5S SIMPT332-6 SIMPT332-6S SIMPT332-6SPR SIMPT332-7 SIMPT332-7S SIMPT332-2SPR SIMPT332-4SPR SIMPT332-5SPR SIMPT332-7SPR
Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Tubes Microcentrifuge Tubes
PP, with O-ring seal screw cap and attachment loop, autoclavable.
  • The O-ring secured in the top of the cap ensures a positive leakproof seal, time after time, keeping the integrity of small samples under even the most adverse conditions
  • Sterile tubes are gamma irradiated and available either plain or with printed graduations and white marking area for sample identification
  • Can be autoclaved to +121 °C in upright position with caps loosened
  • Certified RNase-, DNase-, pyrogen- and DNA-free
Ideal for cryogenic work as well as freezer storage, boilingMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable