Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Supplier: Simport Scientific
T335-2 T335-2S T335-5S T335-6 T335-6S T335-6SPR T335-7 T335-7S T335-7SPR T335-5 T335-5SPR T335-2SPR T335-4 T335-4S T335-4SPR T335-2TP T335-6TP T335-7TP T335-4TP T335-5TP T335-4SPRTP T335-4STP T335-5SPRTP T335-5STP T335-6SPRTP T335-6STP T335-7SPRTP T335-7STP T335-2SPRTP T335-2STP
SIMPT335-2EA 0 SGD
SIMPT335-2 SIMPT335-2S SIMPT335-5S SIMPT335-6 SIMPT335-6S SIMPT335-6SPR SIMPT335-7 SIMPT335-7S SIMPT335-7SPR SIMPT335-5 SIMPT335-5SPR SIMPT335-2SPR SIMPT335-4 SIMPT335-4S SIMPT335-4SPR SIMPT335-2TP SIMPT335-6TP SIMPT335-7TP SIMPT335-4TP SIMPT335-5TP SIMPT335-4SPRTP SIMPT335-4STP SIMPT335-5SPRTP SIMPT335-5STP SIMPT335-6SPRTP SIMPT335-6STP SIMPT335-7SPRTP SIMPT335-7STP SIMPT335-2SPRTP SIMPT335-2STP
Microcentrifuge tubes, 0,5/1,5/1,8 ml, Micrewtube®
Tubes Microcentrifuge Tubes
Tubes and caps made of PP, translucent.
  • Available in plain or graduated configuration
  • Certified RNase, DNase, DNA and pyrogen free
  • Max. RCF: Non skirted tubes can be centrifuged up to 20000×g
  • Temperature range: –196 to +121 °C
  • Flat screw caps with O-ring seal