Culture tubes, sterile, Cultubes™

Supplier: Simport Scientific
T405-1 T405-33 T405-1A T406-1 T406-33 T406-1A T405-2 T405-2A T405-3 T405-6 T405-6A T406-2 T406-2A T406-3 T406-6 T406-6A
SIMPT405-1EA 324 SGD
SIMPT405-1 SIMPT405-33 SIMPT405-1A SIMPT406-1 SIMPT406-33 SIMPT406-1A SIMPT405-2 SIMPT405-2A SIMPT405-3 SIMPT405-6 SIMPT405-6A SIMPT406-2 SIMPT406-2A SIMPT406-3 SIMPT406-6 SIMPT406-6A
Culture tubes, sterile, Cultubes™
Tubes Culture Tubes
PP or PS, graduated, disposable, sterile.

  • White marking field
  • Gradation distinguishing 0,1 (4 ml tube) and 0,25 ml (12 ml tube)
  • Biological inert
  • Durable, reduces the risk in handling potentially hazardous material

Can be used for most routine laboratory procedures.

Resist temperatures from −190 to +120 °C (PP) and up to 70 °C (PS).

Max. RCF: 3000×g (PP); 1400×g (PS).

Delivery information: With or without cap available.
Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR