Centrifuge tubes, conical bottom
Supplier: Simport Scientific
T408-1 T408-2 T408
SIMPT408-1EA 0 SGD
SIMPT408-1 SIMPT408-2 SIMPT408
Centrifuge tubes, conical bottom
Tubes Centrifuge Tubes
PP or PS, conical bottom, graduated, disposable.
  • Suitable for general centrifugation, urinalysis procedures and serum separation
  • Chemically clean and metal free, ready to use and uniform in size and shape
  • Graduated
Max. RCF: 3000×g for PP tubes, 1200×g for PS tubes.
Order Now
SPECIFICATIONS
Environmentally Preferable