Sample containers, with screw cap, non-pyrogenic, Sterilin®
Supplier: Sterilin
128PYR 125PYR 129PYR 190PYR 185PYR
SLIN128PYREA 0 SGD
SLIN128PYR SLIN125PYR SLIN129PYR SLIN190PYR SLIN185PYR
Sample containers, with screw cap, non-pyrogenic, Sterilin®
Sample Containers General Purpose Containers
PS, with PE, PP oder metal screw cap.
  • In vitro use only
  • Gamma irradiated
  • Leak tested in accordance with EN14254 Annex D and BS5213
  • Lot number on each container label and test certificate supplied for each lot
Suitable for the storage of samples/formulation work/ sterility testing and endotoxin testing. Certified endotoxin free to levels below 0,01 EU/ml (Limulus Amoebocyte Lysate (LAL) test).
Perform sterility and endotoxin tests with Thermo Scientific™ Sterilin™ non-pyrogenic polystyrene containers. Not available in America.
Caution: *TheMore Product Information