Sterilin® Dippas® Milk Dipper
Supplier: Sterilin
194IW
SLIN194IWEA 0 SGD
SLIN194IW
Sterilin® Dippas® Milk Dipper
Samplers Sample Dippers
Clear polystyrene container with snap-off handle and leakproof metal screw cap enabling samples to be sent for analysis without risk of cross-contamination.
  • Individually wrapped and sterilized by gamma irradiation
  • Leak tested in accordance with BS EN ISO 6717
  • Available in either clear or blue polystyrene
  • Handle neatly snaps off after sample collection for ease of transportation
  • Choice of handle length to accommodate the majority of sampling applications
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Order Now
SPECIFICATIONS
Environmentally Preferable