Serological pipettes, Sterilin®
Supplier: Sterilin
47110
SLIN47110EA 699 SGD
SLIN47110
Serological pipettes, Sterilin®
Pipettes Serological Pipettes
PS, transparent, disposable, sterile.
  • Clear black printed graduations for maximum clarity, colour coded for easy identification, printed with product code/lot number and expiry date
  • Negative graduations for extra capacity
  • Gamma irradiated to ensure sterility and certified both non pyrogenic (<0,1 EU/ml) and non haemolytic
Ordering information: Batch specific certificate supplied in each case.
Caution: *Without suction adapter
Order Now
SPECIFICATIONS
Environmentally Preferable