Sterilin® Petri Dishes
Supplier: Sterilin
501V
SLIN501VEA 0 SGD
SLIN501V
Sterilin® Petri Dishes
Culture Dishes Petri Dishes
Ideal for applications where a large surface area and very flat base are required.
  • Easy grip ridges on the base to aid individual dish handling
  • Triple vented to aid gaseous exchange
  • Ideal for use in automatic plate pourers
Dimensions shown are for outer Ø base and outer height (mm).
Certifications: Produced and tested in accordance with EN ISO 24998
Order Now
SPECIFICATIONS
Environmentally Preferable