Contact plate, Sterilin®
Supplier: Sterilin
504
SLIN504EA 604 SGD
SLIN504
Contact plate, Sterilin®
Culture Dishes Petri Dishes
Ideal for use in routine hygiene monitoring of surfaces.
  • Concave profile of the dish raises the profile of the media when set giving better contact between the agar bed and surface under test
  • Aseptically manufactured under cleanroom conditions (class 7 ISO 14644) to exclude microbiological contamination
  • Triple vented to aid gaseous exchange
  • Numbered grid on the base facilitates colony counting
  • Deep skirted base aids stability when stacked
Dimensions shown are for outer Ø base and outer height (mm).
Certifications: Conforms to IES 1993 and IPF 1990 standards.