Bags, autoclave, 121 °C, Sterilin®
Supplier: Sterilin
509-17 511-17 510 510L
SLIN509-17EA 0 SGD
SLIN509-17 SLIN511-17 SLIN510 SLIN510L
Bags, autoclave, 121 °C, Sterilin®
Bags Waste Disposal Bags
Specifically designed for contaminated waste disposal in autoclaves or incinerators. Strong, HDPE, white, with blue 'Biohazard' printing.
  • Suitable for high temperature sterilisation at 121 °C
  • Convenient tissue box style cartons dispense bags individually
Polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135 °C. Coated wire stands or cardboard holders available for use with autoclave bags.
Order Now
SPECIFICATIONS
Environmentally Preferable