Sterilin™ Biohazard Bags

Supplier: Sterilin
509LHT 510LHT 510HT 511HT
SLIN509LHTEA 0 SGD
SLIN509LHT SLIN510LHT SLIN510HT SLIN511HT
Sterilin™ Biohazard Bags
Bags Waste Disposal Bags
Sterilin autoclave bags are strong disposable bags with blue biohazard printing specifically designed for contaminated waste disposal in autoclaves or incinerators.

  • HDPE bags are suitable for sterilisation at 121 °C
  • Polypropylene bags are suitable for the inactivation of particularly resistant biological waste at 135 °C
  • Specifically designed for contaminated waste disposal in autoclaves or incinerators
  • Convenient tissue box cartons dispense bags individually
  • Coated wire stands or cardboard holders available for use with autoclave bags
Order Now

SPECIFICATIONS

Environmentally Preferable
Max. temperature 135 ℃

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR