Sterilin® Microtitre Plates, 96 Well
Supplier: Sterilin
612U96 612V96 612F96 611U96 611V96 611F96
SLIN612U96EA 0 SGD
SLIN612U96 SLIN612V96 SLIN612F96 SLIN611U96 SLIN611V96 SLIN611F96
Sterilin® Microtitre Plates, 96 Well
Microplates HTS Microplates
PS, non treated. Used for serology and microbiology. Compatible with automated plate readers and washers.
  • Suitable for use at a wavelength of 340 nm
  • Condensation rings on the lid (supplied separately) together with raised well rims on the base help minimise the risk of contamination from surrounding wells
  • Frosted write-on area on end wall, orientation corners and alpha numeric labelling ensure easy sample identification
  • Hanging well design ensures even temperature distribution around each well
  • Manufactured from virgin polystyrene for excellent optical
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable