Sterilin® Lids for 96-well Microtitre Plates
Supplier: Sterilin
642000
SLIN642000EA 164 SGD
SLIN642000
Sterilin® Lids for 96-well Microtitre Plates
Lids Microplate Lids
Store and transport samples or perform microbiology screenings, EIA/absorbance assays and serology work with 96-well Thermo Scientific™ Sterilin™ Clear Microtiter™ Plates.
  • Manufactured from virgin polystyrene for excellent optical clarity
  • Manufactured in ISO Class 7 Clean Room without surface treatment for a medium binding surface
  • Available irradiated for sterility and a more hydrophilic surface, providing high binding
  • Suitable for use at a wavelength of 340 nm
  • Minimizes the risk of contamination from surrounding wells with condensation rings on the lid
More Product Information