Sterilin™ Test Tubes
Supplier: Sterilin
RT15
SLINRT15EA 0 SGD
SLINRT15
Sterilin™ Test Tubes
Tubes Test Tubes
Avoid the hazards of glass with this safe, convenient alternative. Thermo Scientific™ Sterilin™ Plastic round base tubes are manufactured from virgin polystyrene and supplied non sterile. Not available in all geographic regions.
  • Convenient alternative to glass tubes
  • Suitable for samples ranging from 0,6 to 11 ml
Delivery information: Supplied clean and dust free.