Evacuable pellet dies, Atlas™
Supplier: SPECAC
GS03000
SPCCGS03000EA 1770 SGD
SPCCGS03000
Evacuable pellet dies, Atlas™
Presses
These evacuable pellet dies are designed for compacting powdered samples into discs or briquettes.
  • Can be used to produce sample pellets from 5 to 40 mm diameter
  • Provides pellets with clarity and quality
  • Can cover a range of load capabilities
  • An evacuation port is provided to remove the moisture during pressing process
Suited for the preparation of solid KBr pellets for FT-IR molecular spectroscopic analysis and XRF pellets for X-Ray fluorescence atomic spectroscopic analysis.
Delivery information: Supplied with evacuable base, body, plunger, aMore Product Information