Petrolatum, white USP

Supplier: Spectrum Chemical

PE125-500GM
SPCMPE125-500GMEA 0 SGD
SPCMPE125-500GM
Petrolatum, white USP
Petrolatum

Petrolatum, White, USP is used as a moisturizer to treat or prevent dry, rough, scaly, itchy skin and minor skin irritations. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Storage Temperature: Ambient
MDL Number: MFCD00147839
CAS Number: 8009-03-8

Order Now

Specification Test Results

BHT As Reported
Tocopherols As Reported
Specific Gravity @60 C 0.815 - 0.880
Melting Range 38 - 60 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR