Sodium alginate NF

Supplier: Spectrum Chemical

Synonyms: Alginic acid sodium salt, Algin

S1118-2.5KG
SPCMS1118-2.5KGEA 0 SGD
SPCMS1118-2.5KG
Sodium alginate NF
Sodium alginate

Sodium Alginate, NF is a gum that has no discernible flavor and is used in the food and pharmaceutical industries as an emulsifier and to increase viscosity. All Spectrum Chemical manufactured NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
MDL Number: MFCD00081310
CAS Number: 9005-38-3

Order Now

Specification Test Results

Assay 90.8% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR