Vegetable oil brominated FCC

Supplier: Spectrum Chemical

V1031-500MLGL
SPCMV1031-500MLGLEA 0 SGD
SPCMV1031-500MLGL
Vegetable oil brominated FCC
Vegetable oil brominated

Brominated Vegetable Oil, FCC, also known as BVO, is a mix of plant-derived triglycerides and is used to help emulsify citrus flavored drinks. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Storage Temperature: Ambient
CAS Number: 8016-94-2

Order Now

Specification Test Results

Free Bromine To Pass Test
Specific Gravity 1.325-1.335
Free Fatty Acids (As Oleic Acid) max. 2.5 %
Iodine Value max. 16
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR