Xanthan gum NF

Supplier: Spectrum Chemical

XA105-100GM
SPCMXA105-100GMEA 0 SGD
SPCMXA105-100GM
Xanthan gum NF
Xanthan gum

Xanthan Gum, NF is a natural gum polysaccharide used as a thickener or rheology modifier and emulsion stabilizer. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Density: 1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131256
CAS Number: 11138-66-2
EINECS: 234-394-2

Order Now

Specification Test Results

Assay 91% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR