HF perfusion filters, CultureGard®
Supplier: Spectrum Laboratories
CU2M20512N
SPECCU2M20512NEA 3220 SGD
SPECCU2M20512N
HF perfusion filters, CultureGard®
Filters In-Line Filters
CultureGard® HF perfusion hollow fibre sterilising grade filter for cell culture perfusion. Made with 0,2 µm MWCO DynaFibre® membrane that permits the flow of culture medium while preventing the introduction of foreign microbials.
  • These specialised filters have a large surface area for faster flow rates under lower pressures
  • Lower pressure during filtration reduces the risk of the membrane clogging or rupturing
  • The DynaFibre® membrane is hydrophilic so there is minimum hold-up volume
  • PP vent fibre prevents air lock
  • Inlet/outlet connection: Female luer-Lok®
More Product Information