Cole-Parmer HP-200 Large-Capacity Infrared Hot Plate
Supplier: Cole Parmer
04807-84
STUA04807-84EA 0 SGD
STUA04807-84
Cole-Parmer HP-200 Large-Capacity Infrared Hot Plate
Heaters Hotplates
Using a very efficient infra red heater of just 900 W power, this hotplate with a ceramic plate will boil one litre of water over 30% faster than a conventional ceramic hotplate of 1200 W. Ideal for heating large volumes of liquid.
  • Very efficient heating, saving time and energy
  • Chemically resistant ceramic top
  • 'Hot' warning light for user safety
  • BioCote® antimicrobial protection