Magnetic hotplate stirrers, high capacity, CB302 and SB302
Supplier: Cole Parmer
CB302
STUACB302EA 3350 SGD
STUACB302
Magnetic hotplate stirrers, high capacity, CB302 and SB302
Stirrers Magnetic Hotplate Stirrers
The 'hot' warning light will flash whenever the plate temperature is above 50 °C and will operate even when the hotplate is turned off if the unit is still connected to the mains. Powerful magnets and motor give stirring speed up to 1500 min⁻¹ and volumes up to 15 L. Model SB302 has a robust aluminium/silicon alloy top plate that will withstand the knocks of everyday use, the 600 W element and good heat transmission of this material gives rapid heating and ensures even temperature distribution across the whole of the plate. Model CB302 has an easy to clean glass ceramic top withMore Product Information