QlickSMART SNAPIT Lite Personal Regular Ampoule Openers
Supplier: SWANN MORTON
SN-02RPK
SWANSN-02RPKEA 57.7 SGD
SWANSN-02RPK
QlickSMART SNAPIT Lite Personal Regular Ampoule Openers
Ampoule Openers
Qlicksmart SnapIT is a simply safer way to open glass ampoules and avoid painful and dangerous sharps injuries. This easy-to-use multi-use ampoule opener avoids the unacceptable sharps injury rates seen when ampoules are opened by hand. L×W: 80×13 mm, 11 g. Plastic (body), Silicon (O-ring), Stainless steel (keyring and spring).
  • With SnapIT you can safely:
  • Insert the ampoule lid into your multi-use SnapIT
  • Snap open the ampoule
  • Eject the ampoule lid directly into a sharps container
  • Not autoclavable
There are both personal and trolley versions availableMore Product Information