Methylated spirit (denatured ethanol)

Supplier: TACT CHEMIE S.E.A.
Danger

TMET-140
TACTTMET-140EA 0 SGD
TACTTMET-140
Methylated spirit (denatured ethanol)
Methylated Spirit
Density: 0.81 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR