Hypodermic needles, Agani™
Supplier: TERUMO
AN-2325R1 AN-2332R1 AN-2425R1 AN-2516R1 AN-2525R1 AN-2613R1 AN-2716R1 AN-3013R1 AN-1838R1 AN-1938R1 AN-2025R1 AN-2038R1 AN-2125R1 AN-2138R1 AN-2232R1 AN-2238R1
TERUAN-2325R1EA 17.5 SGD
TERUAN-2325R1 TERUAN-2332R1 TERUAN-2425R1 TERUAN-2516R1 TERUAN-2525R1 TERUAN-2613R1 TERUAN-2716R1 TERUAN-3013R1 TERUAN-1838R1 TERUAN-1938R1 TERUAN-2025R1 TERUAN-2038R1 TERUAN-2125R1 TERUAN-2138R1 TERUAN-2232R1 TERUAN-2238R1
Hypodermic needles, Agani™
Syringe Needles
These needles with thin wall are colour coded according to ISO.
  • Transparent needle hub for quick observation of blood reflux
  • Sterilised with ethylene oxide
  • Latex-free
  • Disposable
Packaging: Individual sterile blister packs with colour coding.
Order Now
SPECIFICATIONS
Environmentally Preferable