Intravenous catheters, Surflash™
Supplier: TERUMO
SR-FF1651
TERUSR-FF1651EA 157 SGD
TERUSR-FF1651
Intravenous catheters, Surflash™
Cannulae
Surflash is made of PU, which is firm at room temperature but soft at body temperature, making the catheter more comfortable.
  • Only exists in the straight version
  • Enables quick and easy confirmation of the placement in the vein by an additional flashback
  • Higher certainty of proper placement
Helpful with patients whose veins are hard to detect and in which venipuncture is more difficult, such as small children, people undergoing chemotherapy.