Infrared printer
Supplier: Testo
0554.0549
TEST0554.0549EA 852 SGD
TEST0554.0549
Infrared printer
Printers Document Printers
The universal IR printer completes data transfers in two seconds. This printer allows users to evaluate restoration measures of air moisture, material equilibrium moisture, and pressure dew point quickly and without error.
  • Hands-free operation
  • Wireless infrared interface for fast data transfer
  • Provides fast, easy to read printouts of measurements
  • Energy saving through auto-off feature
  • Instrument memory for up to 10000 readings
Delivery information: Printer supplied wih a wireless infrared interface, one roll of thermal paper, and four AA batteries.