Digital barometer, 511
Supplier: Testo
0560.0511
TEST0560.0511EA 619 SGD
TEST0560.0511
Digital barometer, 511
Barometers
Pocket absolute pressure meter. Very handy, small and easy to use, this barometer permits measurements of barometric air pressure by entering on-site height above sea level and barometric elevation measurement between two points. Backlit display completes the instrument.
  • Automatic off function
  • Eight switchable pressure units: hPa, mbar, Pa, mmH₂O, mmHg, inH₂O, inHg, psi
  • Barometric altitude measurement
Delivery information: Supplied with protective cap, wrist strap, belt holder and two alkaline AAA batteries for about 500 hours of use without display illumination.