Digital mini surface thermometer
Supplier: Testo
0560.1109
TEST0560.1109EA 120 SGD
TEST0560.1109
Digital mini surface thermometer
Thermometers Digital Thermometers
The mini surface thermometer is used for the most varied methods of surface temperature measurements (for temperature checks in heating engineering, the sanitation sector, or air conditioning/refrigeration and ventilation technology).
  • Universally applicable for surface measurement
  • Large, easy-to-read display
  • Battery status display and easy battery replacement
  • Stainless steel surface temperature probe
  • Auto power-off (1 hour) with low battery display
  • Selectable °C/°F display
Silicon thermal paste can be used for fast measurement results which providesMore Product Information