Digital mini penetration thermometer
Supplier: Testo
0560.1110
TEST0560.1110EA 77.4 SGD
TEST0560.1110
Digital mini penetration thermometer
Thermometers Digital Thermometers
This mini penetration thermometer is used for measuring liquid temperatures in the laboratory, or air temperatures in heating and ventilation technology. It can also be used as a food thermometer. Its penetration probe is excellent for measuring the core temperature of food products during food controls.
  • Large, easy to read display
  • Battery status display and easy battery replacement
Delivery information: Supplied with protective sleeve for probe shaft, easily legible display, batteries and certificate of conformity.